Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Caryophyllene 80 fCC
Catalog No. 502902865
Encompass
-Caryophyllene is a volatile sesquiterpene compound that can be used as a flavoring agent. It is one of the major volatile organic compounds innbspEugenia caryophyllata essential oil Psidium guajava L. leaves Arabidopsis thaliana flowers and Artemisia annua L. aerial parts.
Catalog No. 50-290-2865 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W22520720KGK
-Caryophyllene is a volatile sesquiterpene compound that can be used as a flavoring agent. It is one of the major volatile organic compounds innbspEugenia caryophyllata essential oil Psidium guajava L. leaves Arabidopsis thaliana flowers and Artemisia annua L. aerial parts.