Learn More
Medchemexpress LLC Beta-casein phosphopeptide | 183582-09-4 | 99.19% | 2061.96 | 10 MG
SDP
Catalog No. 5000398346
Encompass_Preferred
beta-Casein phosphopeptide is a biologically active peptide derived from bovine beta-Casein. This monophosphopeptide (phospho-Serine) is primarily used for the characterization of affinity-purified phosphorylated peptides in liquid chromatography and for the analysis or detection of phosphorylated peptides in mass spectrometry. This product is for research use only.
  • Biologically active peptide
  • Used for characterization of phosphorylated peptides
  • Suitable for liquid chromatography and mass spectrometry applications
  • For research use only
Catalog No. 50-003-98346 Supplier Medchemexpress LLC Supplier No. HYP553110MG
beta-Casein phosphopeptide is a biologically active peptide derived from bovine beta-Casein. This monophosphopeptide (phospho-Serine) is primarily used for the characterization of affinity-purified phosphorylated peptides in liquid chromatography and for the analysis or detection of phosphorylated peptides in mass spectrometry. This product is for research use only.
  • Biologically active peptide
  • Used for characterization of phosphorylated peptides
  • Suitable for liquid chromatography and mass spectrometry applications
  • For research use only