Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC beta Catenin Antibody (YA936) | 100 UL
SDP

Catalog No. 5000442947
Encompass_Preferred

beta Catenin Antibody (YA936) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to beta Catenin. It is for research use only. The antibody is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, with a pH of 7.3. It should be stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles, and shipped with blue ice. The species reactivity includes Human, Mouse, and Rat.

  • Monoclonal antibody (IgG1)
  • Targets beta Catenin
  • For research use only
  • Suitable for Western Blot (1:500-1:1000)
  • Suitable for Immunohistochemistry-Paraffin (1:50-1:100)
  • Species reactivity: Human, Mouse, Rat
  • Predicted band size: 85 kDa
  • Observed band size: 92 kDa
  • Storage: -20°C for 1 year

Catalog No. 50-004-42947 Supplier Medchemexpress LLC Supplier No. HYP81261100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

beta Catenin Antibody (YA936) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, targeting to beta Catenin. It is for research use only. The antibody is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, with a pH of 7.3. It should be stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles, and shipped with blue ice. The species reactivity includes Human, Mouse, and Rat.

  • Monoclonal antibody (IgG1)
  • Targets beta Catenin
  • For research use only
  • Suitable for Western Blot (1:500-1:1000)
  • Suitable for Immunohistochemistry-Paraffin (1:50-1:100)
  • Species reactivity: Human, Mouse, Rat
  • Predicted band size: 85 kDa
  • Observed band size: 92 kDa
  • Storage: -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.