Learn More
Medchemexpress LLC beta-Catenin Human 5ug
SDP
Catalog No. 5000178833
Encompass_Preferred
Beta-Catenin protein is an important component of the canonical Wnt pathway and regulates cellular processes In the absence of Wnt it complexes with AXIN1 AXIN2 APC CSNK1A1 and GSK3B to promote CTNNB1 phosphorylation and degradation beta-Catenin Protein Human (sf9 His-GST) is the recombinant human-derived beta-Catenin protein expressed by Sf9 insect cells with N-His N-GST labeled tag
Catalog No. 50-001-78833 Supplier Medchemexpress LLC Supplier No. HYP743825UG
Beta-Catenin protein is an important component of the canonical Wnt pathway and regulates cellular processes In the absence of Wnt it complexes with AXIN1 AXIN2 APC CSNK1A1 and GSK3B to promote CTNNB1 phosphorylation and degradation beta-Catenin Protein Human (sf9 His-GST) is the recombinant human-derived beta-Catenin protein expressed by Sf9 insect cells with N-His N-GST labeled tag