Learn More
Medchemexpress LLC Beta-catenin-IN-2 | 1458664-10-2 | 99.55% | 255.29 | 100 MG
SDP
Catalog No. 5000350804
Encompass_Preferred
Beta-catenin-IN-2 (Compound H1B1) is a potent β-catenin inhibitor used for the study of colorectal cancer. It suppresses the transcriptional activity of β-catenin/Tcf-4 in HCT116 cells in a dose-dependent manner at concentrations of 10-40 μM. The product is a solid, pink to red in color. It is soluble in DMSO at 83.33 mg/mL (326.41 mM).
  • Potent β-catenin inhibitor
  • Suppresses transcriptional activity of β-catenin/Tcf-4
  • Used for the study of colorectal cancer
  • Solid, pink to red in color
  • Soluble in DMSO
Catalog No. 50-003-50804 Supplier Medchemexpress LLC Supplier No. HY136464100MG
Beta-catenin-IN-2 (Compound H1B1) is a potent β-catenin inhibitor used for the study of colorectal cancer. It suppresses the transcriptional activity of β-catenin/Tcf-4 in HCT116 cells in a dose-dependent manner at concentrations of 10-40 μM. The product is a solid, pink to red in color. It is soluble in DMSO at 83.33 mg/mL (326.41 mM).
  • Potent β-catenin inhibitor
  • Suppresses transcriptional activity of β-catenin/Tcf-4
  • Used for the study of colorectal cancer
  • Solid, pink to red in color
  • Soluble in DMSO