Learn More
Enzo Life Sciences beta-Catenin monoclonal antibody (12F7) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMST001E
Encompass_Preferred
Clone: 12F7. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant β-catenin (N-terminal fragment). UniProt ID: P35222 (human), Q02248 (mouse), Q9WU82 (rat). Species reactivity: Human, Mouse, Rat. Applications: IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (1-5µg/ml) Immunoprecipitation (2µg/200-300µl cell extract) Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7257