Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Chlorophenetole 95.0+%, TCI America™
SDP

Catalog No. C024025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C024025ML 25 mL
C0240500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. C024025ML Supplier TCI America Supplier No. C024025ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 622-86-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00018946
InChI Key VQUYNUJARXBNPK-UHFFFAOYSA-N
Synonym 2-chloroethoxy benzene, beta-chlorophenetole, 2-phenoxyethyl chloride, benzene, 2-chloroethoxy, 2-chloroethyl phenyl ether, b-chlorophenetole, phenetole, .beta.-chloro, phenoxyethyl chloride, benzene, chloroethoxy, 2-chloroethyoxy benzene
PubChem CID 12156
IUPAC Name 2-chloroethoxybenzene
SMILES C1=CC=C(C=C1)OCCCl

Specifications

Color White
Boiling Point 220°C
Quantity 25 mL
Formula Weight 156.61
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material beta-Chlorophenetole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.