Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-beta-Citronellol, TCI America™
SDP

Catalog No. C225425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C225425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C225425ML Supplier TCI America Supplier No. C225425ML

missing translation for 'validMainframeMsg'

Specifications

CAS 7540-51-4
Color Colorless
Boiling Point 118°C
Molecular Formula C10H20O
MDL Number MFCD00063214
Quantity 25 mL
Synonym --citronellol, s-3,7-dimethyloct-6-en-1-ol, --beta-citronellol, s---citronellol, beta-rhodinol, s---beta-citronellol, s-citronellol, --3,7-dimethyloct-6-en-1-ol, 3s-3,7-dimethyloct-6-en-1-ol, unii-8rsy5y5658
InChI Key QMVPMAAFGQKVCJ-JTQLQIEISA-N
SMILES CC(CCC=C(C)C)CCO
IUPAC Name (3S)-3,7-dimethyloct-6-en-1-ol
Molecular Weight (g/mol) 156.269
PubChem CID 7793
ChEBI CHEBI:88
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-beta-Citronellol
Show More Show Less
EINECSNumber (2)-0258
RTECSNumber RH3400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.