Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-cyclodextrin | 7585-39-9 | MFCD00078139 | 100.0% | 1134.98 | C42H70O35 | 500 G
SDP

Catalog No. 5000240429
Encompass_Preferred

β-Cyclodextrin is a cyclic oligosaccharide of seven glucose units with a hydrophobic central cavity and a hydrophilic exterior. It is commonly used in research to enhance solubility, stabilize and encapsulate guest molecules, and support formulation and drug-delivery studies.

  • Cyclic heptasaccharide structure with a hydrophobic central cavity.
  • Molecular formula C42H70O35.
  • Molecular weight 1134.98.
  • High purity (≈99.95%).
  • White crystalline powder physical form.
  • Applications include solubilization, encapsulation, stabilization, and drug-delivery research.
  • Available in laboratory-scale pack sizes including 500 g.

Catalog No. 50-002-40429 Supplier Medchemexpress LLC Supplier No. HY107201500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

β-Cyclodextrin is a cyclic oligosaccharide of seven glucose units with a hydrophobic central cavity and a hydrophilic exterior. It is commonly used in research to enhance solubility, stabilize and encapsulate guest molecules, and support formulation and drug-delivery studies.

  • Cyclic heptasaccharide structure with a hydrophobic central cavity.
  • Molecular formula C42H70O35.
  • Molecular weight 1134.98.
  • High purity (≈99.95%).
  • White crystalline powder physical form.
  • Applications include solubilization, encapsulation, stabilization, and drug-delivery research.
  • Available in laboratory-scale pack sizes including 500 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.