Learn More
Medchemexpress LLC Beta-cyclodextrin | 7585-39-9 | MFCD00078139 | 100.0% | 1134.98 g/mol | C42H70O35 | 10 G
SDP
Catalog No. 5000240930
Encompass_Preferred
β-Cyclodextrin is a cyclic oligosaccharide made of seven α-D-glucopyranose units that creates a toroidal structure with a hydrophobic cavity and hydrophilic exterior. Supplied as a white to off-white powder for research and formulation use, it is commonly employed to improve aqueous solubility, stability, and bioavailability of poorly soluble compounds.
  • Cyclic oligosaccharide composed of seven glucose units.
  • Forms inclusion complexes to enhance solubility and stability.
  • Molecular weight approximately 1134.98 g/mol.
  • High purity suitable for research applications.
  • White to off-white solid powder.
  • Used in pharmaceutical formulation and analytical studies.
  • Store powder under recommended cold conditions for long-term stability.
Catalog No. 50-002-40930 Supplier Medchemexpress LLC Supplier No. HY10720110G
β-Cyclodextrin is a cyclic oligosaccharide made of seven α-D-glucopyranose units that creates a toroidal structure with a hydrophobic cavity and hydrophilic exterior. Supplied as a white to off-white powder for research and formulation use, it is commonly employed to improve aqueous solubility, stability, and bioavailability of poorly soluble compounds.
  • Cyclic oligosaccharide composed of seven glucose units.
  • Forms inclusion complexes to enhance solubility and stability.
  • Molecular weight approximately 1134.98 g/mol.
  • High purity suitable for research applications.
  • White to off-white solid powder.
  • Used in pharmaceutical formulation and analytical studies.
  • Store powder under recommended cold conditions for long-term stability.