Learn More
Medchemexpress LLC Beta cyclodextrin | 7585-39-9 | MFCD00078139 | >=95.0% | 1,134.98 g/mol | C42H70O35 | 25 G
SDP
Catalog No. 5000241068
Encompass_Preferred
β-Cyclodextrin is a cyclic heptamer of glucose used to form inclusion complexes that increase the solubility, stability, and bioavailability of hydrophobic guest molecules. Supplied as a dry powder for laboratory and formulation work, it is commonly used in drug delivery research, analytical sample preparation, and excipient studies.
  • Improves solubility of hydrophobic compounds
  • Forms inclusion complexes with a wide range of guest molecules
  • Supports drug delivery and formulation development studies
  • Useful for analytical sample preparation and chromatography
  • Supplied as a stable dry powder for easy handling and storage
Catalog No. 50-002-41068 Supplier Medchemexpress LLC Supplier No. HY10720125G
β-Cyclodextrin is a cyclic heptamer of glucose used to form inclusion complexes that increase the solubility, stability, and bioavailability of hydrophobic guest molecules. Supplied as a dry powder for laboratory and formulation work, it is commonly used in drug delivery research, analytical sample preparation, and excipient studies.
  • Improves solubility of hydrophobic compounds
  • Forms inclusion complexes with a wide range of guest molecules
  • Supports drug delivery and formulation development studies
  • Useful for analytical sample preparation and chromatography
  • Supplied as a stable dry powder for easy handling and storage