Learn More
Selleck Chemical LLC Beta-Cyclodextrin-E0046-1G
SDP
Catalog No. 503135254
Encompass_Preferred
-Cyclodextrin ( -CD Beta-Cyclodextrin) is a cyclic derivative of starch prepared from partially hydrolyzed starch (maltodextrin) by an enzymatic process -Cyclodextrin can be used as complexing agents to increase aqueous solubility of poorly soluble drugs and to increase their bioavailability and stability
Catalog No. 50-313-5254 Supplier Selleck Chemical LLC Supplier No. E00461G
-Cyclodextrin ( -CD Beta-Cyclodextrin) is a cyclic derivative of starch prepared from partially hydrolyzed starch (maltodextrin) by an enzymatic process -Cyclodextrin can be used as complexing agents to increase aqueous solubility of poorly soluble drugs and to increase their bioavailability and stability