Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Cyclodextrin polymer5G
Catalog No. 502905822
Encompass
Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via(1 4) glycosidic bonds. Common cyclodextrins contain six (CD) seven (-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivatized through esterification at positions two three and/or six.
Catalog No. 50-290-5822 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C24855G
Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via(1 4) glycosidic bonds. Common cyclodextrins contain six (CD) seven (-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivatized through esterification at positions two three and/or six.