Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Beta-D-Galactose Pentaacetate

Catalog No. 134031100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-403-1100G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-403-1100G Supplier Sigma Aldrich Supplier No. 134031100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Beta-D-Galactose Pentaacetate

Specifications

CAS 105379-24-6
Melting Point 176°C
Molecular Formula C15H20F6N5OP
Linear Formula C15H20F6N5OP
MDL Number MFCD00191770
Quantity 25 g
Synonym 1,2,3,4,6-Penta-O-acetyl-beta-D-galactopyranose; Penta-O-acetyl-beta-D-galactopyranose
Molecular Weight (g/mol) 431.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.