Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-D-galactose pentaacetate | 4163-60-4 | MFCD00063259 | 99.6% | 390.34 | C16H22O11 | 1000 G
SDP

Catalog No. 5000299493
Encompass_Preferred

β-D-Galactose pentaacetate is the peracetylated derivative of beta-D-galactose used as a protected sugar reagent in organic synthesis and biochemical research. Supplied as a high-purity solid for research use.

  • Peracetylated monosaccharide useful for glycosylation and derivatization reactions.
  • Supplied as a solid with high purity suitable for research applications.
  • Molecular formula C16H22O11 and molecular weight 390.34 g/mol.
  • Storage: solid under nitrogen at 4°C; in solution store short-term at -20°C to -80°C.
  • Available in multiple pack sizes up to 1000 g.

Catalog No. 50-002-99493 Supplier Medchemexpress LLC Supplier No. HYW0399231000G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Galactose pentaacetate is the peracetylated derivative of beta-D-galactose used as a protected sugar reagent in organic synthesis and biochemical research. Supplied as a high-purity solid for research use.

  • Peracetylated monosaccharide useful for glycosylation and derivatization reactions.
  • Supplied as a solid with high purity suitable for research applications.
  • Molecular formula C16H22O11 and molecular weight 390.34 g/mol.
  • Storage: solid under nitrogen at 4°C; in solution store short-term at -20°C to -80°C.
  • Available in multiple pack sizes up to 1000 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.