Learn More
Medchemexpress LLC Beta-D-galactose pentaacetate | 4163-60-4 | MFCD00063259 | 99.6% | 390.34 g/mol | C16H22O11 | 1 ML
SDP
Catalog No. 5000300223
Encompass_Preferred
β-D-Galactose pentaacetate is the penta-acetylated derivative of D-galactose used as a biochemical research reagent. It acts as an insulin antagonist and is hydrolyzed by intracellular esterases. The compound is supplied as a solid and as ready-to-use 10 mM solutions for laboratory assays.
  • High assay purity (~99.6%) for consistent experimental results.
  • Available as 10 mM solution in DMSO for immediate use in assays.
  • White to off-white solid with long shelf stability when stored properly.
  • Chemical formula C16H22O11; molecular weight 390.34 g/mol.
  • Store solid at 4°C; solutions stable at -20°C to -80°C under inert atmosphere.
Catalog No. 50-003-00223 Supplier Medchemexpress LLC Supplier No. HYW03992310MM/1ML
β-D-Galactose pentaacetate is the penta-acetylated derivative of D-galactose used as a biochemical research reagent. It acts as an insulin antagonist and is hydrolyzed by intracellular esterases. The compound is supplied as a solid and as ready-to-use 10 mM solutions for laboratory assays.
  • High assay purity (~99.6%) for consistent experimental results.
  • Available as 10 mM solution in DMSO for immediate use in assays.
  • White to off-white solid with long shelf stability when stored properly.
  • Chemical formula C16H22O11; molecular weight 390.34 g/mol.
  • Store solid at 4°C; solutions stable at -20°C to -80°C under inert atmosphere.