Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-D-glucopyranose | 492-61-5 | 99.9% | 1 G
SDP

Catalog No. 5000230545
Encompass_Preferred

β-D-Glucose (beta-D-glucopyranose) is a naturally occurring monosaccharide used as a biochemical reagent and endogenous metabolite in research applications. It is supplied as a solid suitable for assay development, cell culture supplementation, and analytical workflows.

  • High purity, suitable for research applications.
  • Molecular formula C6H12O6 and molecular weight 180.16 g/mol.
  • White to light yellow solid, stable under recommended storage.
  • Store at 4°C; in solution store at -20°C (1 month) or -80°C (6 months).
  • Available in small pack sizes for laboratory use (1 G).
  • Documentation includes data sheet, certificate of analysis, and SDS.

Catalog No. 50-002-30545 Supplier Medchemexpress LLC Supplier No. HY1219651G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-D-Glucose (beta-D-glucopyranose) is a naturally occurring monosaccharide used as a biochemical reagent and endogenous metabolite in research applications. It is supplied as a solid suitable for assay development, cell culture supplementation, and analytical workflows.

  • High purity, suitable for research applications.
  • Molecular formula C6H12O6 and molecular weight 180.16 g/mol.
  • White to light yellow solid, stable under recommended storage.
  • Store at 4°C; in solution store at -20°C (1 month) or -80°C (6 months).
  • Available in small pack sizes for laboratory use (1 G).
  • Documentation includes data sheet, certificate of analysis, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.