Learn More
Medchemexpress LLC Beta-D-glucuronide-pNP-carbonate | 894095-98-8 | MFCD31715405 | >96.0% | 913.83 g/mol | C45H43N3O18 | 50 MG
SDP
Catalog No. 5000203877
Encompass_Preferred
β-D-glucuronide-pNP-carbonate is a glycosidase-cleavable antibody-drug conjugate (ADC) linker that contains a para-nitrophenyl carbonate reactive handle and a β-D-glucuronide cleavable trigger. It is supplied as a solid for research use and is designed to release payloads upon enzymatic cleavage by glucuronidase.
  • Glycosidase-cleavable linker for targeted payload release.
  • Para-nitrophenyl carbonate reactive handle for payload attachment.
  • Solid form suitable for standard laboratory handling.
  • Available in small research-scale quantities for synthesis work.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-03877 Supplier Medchemexpress LLC Supplier No. HY13632950MG
β-D-glucuronide-pNP-carbonate is a glycosidase-cleavable antibody-drug conjugate (ADC) linker that contains a para-nitrophenyl carbonate reactive handle and a β-D-glucuronide cleavable trigger. It is supplied as a solid for research use and is designed to release payloads upon enzymatic cleavage by glucuronidase.
  • Glycosidase-cleavable linker for targeted payload release.
  • Para-nitrophenyl carbonate reactive handle for payload attachment.
  • Solid form suitable for standard laboratory handling.
  • Available in small research-scale quantities for synthesis work.
  • Intended for research use only; not for clinical or human use.