Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-D-Lactose, contains ^=80% beta and ^=20% alpha

Catalog No. AAH54447A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54447A1 1 kg
AAH5444718 50 g
AAH5444730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54447A1 Supplier Thermo Scientific Chemicals Supplier No. H54447A1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5965-66-2 | C12H22O11 | 342.30 g/mol

β-D-Lactose is used in the pharmaceutical industry to make tablets. It acts as a nutrient and as a filler used in pills. It is utilized in the dilution of heroin. Further, it is used to sweeten stout beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-D-Lactose is used in the pharmaceutical industry to make tablets. It acts as a nutrient and as a filler used in pills. It is utilized in the dilution of heroin. Further, it is used to sweeten stout beer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-66-2
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00064521
InChI Key GUBGYTABKSRVRQ-DCSYEGIMSA-N
Synonym beta-d-lactose, beta-lactose, lactose, lactose, anhydrous, fast-flo lactose, milk sugar, d-lactose, 4-o-beta-d-galactopyranosyl-beta-d-glucopyranose, fast-flo, lactose anhydrous
PubChem CID 6134
ChEBI CHEBI:36218
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 1 kg
Merck Index 14,5343
Solubility Information Soluble in water.
Formula Weight 342.3
Chemical Name or Material beta-D-Lactose, contains ≈70% beta and ≈30% alpha
EINECSNumber 227-751-9
RTECSNumber LZ5776360
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.