Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-D-Maltose octaacetate, 98%

Catalog No. AAA1298806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1298806 5 g
AAA1298814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1298806 Supplier Thermo Scientific Chemicals Supplier No. A1298806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 22352-19-8 | C28H38O19 | 678.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22352-19-8
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.593
MDL Number MFCD00079432
InChI Key WOTQVEKSRLZRSX-FKNZMNJBSA-N
Synonym Octa-O-acetyl-beta-D-maltose
PubChem CID 126963554
IUPAC Name [(2S,3R,4S,5R,6S)-4,5,6-triacetyloxy-3-[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 157°C to 160°C
Quantity 5 g
Beilstein 79279
Optical Rotation +65° (c=10 in Chloroform)
Formula Weight 678.6
Percent Purity 98%
Chemical Name or Material beta-D-Maltose octaacetate
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.