Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Beta-D-Mannopyranose 1,3,4,6-Tetra-O-Acetate 2-O-Trifluoromethanesulfonate

Catalog No. 655112100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-511-2100MG 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-511-2100MG Supplier Sigma Aldrich Supplier No. 655112100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Beta-D-Mannopyranose 1,3,4,6-Tetra-O-Acetate 2-O-Trifluoromethanesulfonate

Specifications

CAS 63449-41-2
Linear Formula C12H16BNO4
Quantity 500 mL
MDL Number MFCD00145757
Synonym Alkylbenzyldimethylammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.