Learn More
Medchemexpress LLC Beta-D-ribofuranose, 1,2,3,5-tetraacetate | 13035-61-5 | MFCD00005358 | 98.0% | 318.28 g/mol | C13H18O9 | 1 ML
SDP
Catalog No. 5000301630
Encompass_Preferred
Beta-D-ribofuranose, 1,2,3,5-tetraacetate is an acetylated sugar derivative used as a biochemical reagent and synthetic intermediate in nucleoside and nucleotide chemistry. It is supplied in prepared solution form (for example, 10 mM in 1 mL) and as bulk material for research applications.
  • Acetylated derivative of D-ribofuranose.
  • Serves as a precursor in nucleoside and nucleotide synthesis.
  • Available as solution and bulk solid for flexible experimental use.
  • Molecular weight 318.28 g/mol, useful for stoichiometric calculations.
  • Commonly used in organic synthesis and biochemical research workflows.
Catalog No. 50-003-01630 Supplier Medchemexpress LLC Supplier No. HYW01833410MM/1ML
Beta-D-ribofuranose, 1,2,3,5-tetraacetate is an acetylated sugar derivative used as a biochemical reagent and synthetic intermediate in nucleoside and nucleotide chemistry. It is supplied in prepared solution form (for example, 10 mM in 1 mL) and as bulk material for research applications.
  • Acetylated derivative of D-ribofuranose.
  • Serves as a precursor in nucleoside and nucleotide synthesis.
  • Available as solution and bulk solid for flexible experimental use.
  • Molecular weight 318.28 g/mol, useful for stoichiometric calculations.
  • Commonly used in organic synthesis and biochemical research workflows.