Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-D-ribofuranose 1,2,3,5-tetraacetate | 13035-61-5 | MFCD00005358 | 98.0% | 318.28 g·mol⁻¹ | C13H18O9 | 500 G
SDP

Catalog No. 5000299682
Encompass_Preferred

Beta-D-ribofuranose 1,2,3,5-tetraacetate is an acetylated ribose derivative used as a protected sugar intermediate and biochemical reagent in organic synthesis and life-science research. It is supplied as a solid with documented solubility and formulation data for in vitro and in vivo use.

  • Purity 98.0%.
  • Molecular weight 318.28 g·mol⁻¹.
  • Chemical formula C13H18O9.
  • Soluble in DMSO (50 mg/mL; ultrasonic may be required).
  • Supplied as a white to off-white solid; available in 500 g and bulk quantities.

Catalog No. 50-002-99682 Supplier Medchemexpress LLC Supplier No. HYW018334500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta-D-ribofuranose 1,2,3,5-tetraacetate is an acetylated ribose derivative used as a protected sugar intermediate and biochemical reagent in organic synthesis and life-science research. It is supplied as a solid with documented solubility and formulation data for in vitro and in vivo use.

  • Purity 98.0%.
  • Molecular weight 318.28 g·mol⁻¹.
  • Chemical formula C13H18O9.
  • Soluble in DMSO (50 mg/mL; ultrasonic may be required).
  • Supplied as a white to off-white solid; available in 500 g and bulk quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.