Learn More
CPC Scientific H-Asp-His-Tyr-Asn-Cys-Val-Ser-Ser-Gly-Gly-Gln-Cys-Leu-Tyr-Ser-Ala-Cys-Pro-Ile-Phe-Thr-Lys-Ile-Gln-Gly-Thr-Cys-Tyr-Arg-Gly-Lys-Ala-Lys-Cys-Cys-Lys-OH (trifluoroacetate salt) 0.1MG
SDP
Supplier:  CPC Scientific DEFS003A
Encompass
SEQUENCE: H-Asp-His-Tyr-Asn-Cys-Val-Ser-Ser-Gly-Gly-Gln-Cys-Leu-Tyr-Ser-Ala-Cys-Pro-Ile-Phe-Thr-Lys-Ile-Gln-Gly-Thr-Cys-Tyr-Arg-Gly-Lys-Ala-Lys-Cys-Cys-Lys-OH (trifluoroacetate salt)(Cys5 and 34 bridge, Cys12 and 27 bridge, Cys17 and 35 bridge)ONE-LETTER SEQUENCE: DHYNCVSSGGQCLYSACPIFTKIQGTCYRGKAKCCK (C5&C34 bridge, C12&C27 bridge, C17&C35 bridge)MOLECULAR FORMULA: C167H256N48O50S6MOLECULAR WEIGHT: 3928.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: -Defensin-1, humanRESEARCH AREA: AntimicrobialREFERENCES:K.W. Bensch, M. Raida, H-J. M?gert, P. Schulz-Knappe, and W.-G. Forssmann, FEBS Lett., 368, 331 (1995). (Original Article)R.I. Lehrer and T. Ganz, Ann. N.Y. Acad. Sci., 797, 228 (1996). (Review Article)SKU(s): DEFS-003A, DEFS-003B
Catalog No. 50-210-9188