Learn More
CPC Scientific H-Gly-Ile-Gly-Asp-Pro-Val-Thr-Cys-Leu-Lys-Ser-Gly-Ala-Ile-Cys-His-Pro-Val-Phe-Cys-Pro-Arg-Arg-Tyr-Lys-Gln-Ile-Gly-Thr-Cys-Gly-Leu-Pro-Gly-Thr-Lys-Cys-Cys-Lys-Lys-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific DEFS004C
Encompass
ONE-LETTER SEQUENCE: GIGDPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP (C8&C37 bridge, C20&C38 bridge, C15&C30 bridge)MOLECULAR FORMULA: C188H305N55O50S6MOLECULAR WEIGHT:4328.23STORAGE CONDITIONS: -20 5CSYNONYMS: β-Defensin 2 (human), β-Defensin 2 (human)RESEARCH AREA: Antimicrobial
Catalog No. 50-210-9192