Learn More
Enzo Life Sciences beta-Defensin 3 (human), (recombinant) (20µg)
SDP
Catalog No. 502013984
Encompass_Preferred
Alternative name: BD-3. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% trifluoroacetic acid (TFA). Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-3984 Supplier Enzo Life Sciences Supplier No. ENZPRT1970020
Alternative name: BD-3. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% trifluoroacetic acid (TFA). Source: Produced in E. coli. Long Term Storage: -80°C.