Learn More
TARGETMOL CHEMICALS INC BETA-ELEMENE 100MG
SDP
Catalog No. 502581343
Encompass_Preferred
Also available in 20 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Elemene (Levo-(beta)-elemene) is a natural product isolated from Curcuma wenyujin with an antitumor activity and induce cell apoptosis. purity: 99%
Catalog No. 50-258-1343 Supplier TARGETMOL CHEMICALS INC Supplier No. T13476100MG
Also available in 20 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (beta)-Elemene (Levo-(beta)-elemene) is a natural product isolated from Curcuma wenyujin with an antitumor activity and induce cell apoptosis. purity: 99%