Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Endothelial Cell Grow
Catalog No. 502902695
Encompass
The -endothelial cell growth factor (b-ECGf) gene encodes an angiogenic factor and is mapped to human chromosome 11q13.1. Recombinant human b-endothelial cell growth factor (b-ECGf) is a 14 amino acid N-terminal extension of acidic fibroblast growth factor. It is expressed in Escherichia coli. The 154 amino acid recombinant protein generated after removal of the initiation methionine has a predicted molecular mass of about 17 kDa.
Catalog No. 50-290-2695 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E138825UG
The -endothelial cell growth factor (b-ECGf) gene encodes an angiogenic factor and is mapped to human chromosome 11q13.1. Recombinant human b-endothelial cell growth factor (b-ECGf) is a 14 amino acid N-terminal extension of acidic fibroblast growth factor. It is expressed in Escherichia coli. The 154 amino acid recombinant protein generated after removal of the initiation methionine has a predicted molecular mass of about 17 kDa.