Learn More
Medchemexpress LLC Beta-estradiol 17-hemisuccinate | 7698-93-3 | MFCD00056186 | 98.8% | 372.45 g/mol | C22H28O5 | 10 MG
SDP
Catalog No. 5000287121
Encompass_Preferred
β-Estradiol 17-hemisuccinate is a synthetic hemisuccinate derivative of estradiol provided for laboratory research applications. It is supplied as a white to off-white solid with high reported purity and is used in biochemical and pharmacological studies of estrogenic activity. Follow recommended storage and handling to maintain stability and assay performance.
  • Synthetic hemisuccinate derivative of estradiol for research use.
  • White to off-white solid with high reported purity (≈98.8%).
  • Provided in small laboratory quantities suitable for assay work.
  • Store at 4°C under nitrogen; in solvent, store at -80°C for long-term.
  • Applicable to biochemical and pharmacological studies involving estrogenic compounds.
Catalog No. 50-002-87121 Supplier Medchemexpress LLC Supplier No. HY14858610MG
Add to Cart
Add to Cart
β-Estradiol 17-hemisuccinate is a synthetic hemisuccinate derivative of estradiol provided for laboratory research applications. It is supplied as a white to off-white solid with high reported purity and is used in biochemical and pharmacological studies of estrogenic activity. Follow recommended storage and handling to maintain stability and assay performance.
  • Synthetic hemisuccinate derivative of estradiol for research use.
  • White to off-white solid with high reported purity (≈98.8%).
  • Provided in small laboratory quantities suitable for assay work.
  • Store at 4°C under nitrogen; in solvent, store at -80°C for long-term.
  • Applicable to biochemical and pharmacological studies involving estrogenic compounds.