Learn More
Apexbio Technology LLC BETA-EUDESMOL-1MG
SDP
Catalog No. 5000570561
Encompass_Preferred
Beta-Eudesmol (CAS No 473-15-4) is a sesquiterpenoid alcohol commonly researched for its pharmacological properties Originating from plants like Atractylodes lancea it targets various biochemical pathways notably influencing neurological and inflammatory responses Studies indicate that its in vitro activity often falls in the nanomolar to low micromolar range showing potential in modulating cellular processes It is frequently utilized in cell models such as neuronal and cancer cell lines and has been the subject of animal studies to explore its therapeutic potential Beta-Eudesmol is valuable in drug screening due to its diverse biological effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70561 Supplier Apexbio Technology LLC Supplier No. BA33851
Beta-Eudesmol (CAS No 473-15-4) is a sesquiterpenoid alcohol commonly researched for its pharmacological properties Originating from plants like Atractylodes lancea it targets various biochemical pathways notably influencing neurological and inflammatory responses Studies indicate that its in vitro activity often falls in the nanomolar to low micromolar range showing potential in modulating cellular processes It is frequently utilized in cell models such as neuronal and cancer cell lines and has been the subject of animal studies to explore its therapeutic potential Beta-Eudesmol is valuable in drug screening due to its diverse biological effects Experimental concentrations typically depend on the specific research design