Learn More
Biotium Chromogenic beta-galactosidase Substrate sampler kit
Catalog No. 501964099
Encompass_Preferred
For researchers wishing to investigate the suitability of our various chromogenic -galactosidase substrates, this sampler kit provides 5 mg each of the following five different substrates: X-Gal, Red-D-Gal, Rose--D-Gal, Purple--D-Gal, and Green--D-Gal.
Catalog No. 50-196-4099 Supplier Biotium Supplier No. 10024
For researchers wishing to investigate the suitability of our various chromogenic -galactosidase substrates, this sampler kit provides 5 mg each of the following five different substrates: X-Gal, Red-D-Gal, Rose--D-Gal, Purple--D-Gal, and Green--D-Gal.