Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-galactosidase protein, Saccharolobus solfataricus (FLAG, His) | >85.0% | 10UG
SDP

Catalog No. 5000176045
Encompass_Preferred

Recombinant beta-galactosidase protein from Saccharolobus solfataricus, expressed in Escherichia coli and supplied with N-terminal FLAG and 6xHis tags. Supplied as purified protein for research use in small pack sizes, stored at -80 °C; purity greater than 85% by reducing SDS-PAGE and molecular weight ~59.5 kDa.

  • Recombinant protein expressed in Escherichia coli
  • N-terminal FLAG and 6xHis tags for detection and purification
  • Purity >85% as determined by reducing SDS-PAGE
  • Molecular weight approximately 59.5 kDa
  • Store at -80 °C; avoid repeated freeze-thaw cycles

Catalog No. 50-001-76045 Supplier Medchemexpress LLC Supplier No. HYP70189210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant beta-galactosidase protein from Saccharolobus solfataricus, expressed in Escherichia coli and supplied with N-terminal FLAG and 6xHis tags. Supplied as purified protein for research use in small pack sizes, stored at -80 °C; purity greater than 85% by reducing SDS-PAGE and molecular weight ~59.5 kDa.

  • Recombinant protein expressed in Escherichia coli
  • N-terminal FLAG and 6xHis tags for detection and purification
  • Purity >85% as determined by reducing SDS-PAGE
  • Molecular weight approximately 59.5 kDa
  • Store at -80 °C; avoid repeated freeze-thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.