Learn More
Bioworld Beta-Glactosidase, 5000 Unit(s)
Catalog No. 50198961
Encompass_Preferred
Beta-Galactosidase Enzyme found in lac+ bacterial cells that catalyzes the cleavage of -linked terminal galactose-containing substratesLyophilized powderSource: E. coliConjugation gradeActivity (37°C):800-1000U/mg proteinUnit definition: One unit will hydrolyze 1.0 μmole of o-nitrophenyl β-D-galactoside to o-nitrophenol and D-galactose per min at pH7.3 at 37°C, 410nm.Aggregation (HPLC): <3.0%Application: Used as an indicator enzyme conjugated to proteins such as antibodies.
Catalog No. 50-198-961 Supplier Bioworld Supplier No. 215600022
Beta-Galactosidase Enzyme found in lac+ bacterial cells that catalyzes the cleavage of -linked terminal galactose-containing substratesLyophilized powderSource: E. coliConjugation gradeActivity (37°C):800-1000U/mg proteinUnit definition: One unit will hydrolyze 1.0 μmole of o-nitrophenyl β-D-galactoside to o-nitrophenol and D-galactose per min at pH7.3 at 37°C, 410nm.Aggregation (HPLC): <3.0%Application: Used as an indicator enzyme conjugated to proteins such as antibodies.