Learn More
Sigma Aldrich Fine Chemicals Biosciences beta-Galactosidase from E.
Catalog No. 502901864
Encompass
-Galactosidase or -D-Galactoside galactohydrolase is basically a tetramer made up of identical four polypeptide chains each constituting 1023 amino acids. It is very specific for D-galactose and requires K or Na and Mg2 to be fully active. -Galactosidase enzyme with an oxygen glycosidic bond catalyzes reactions with -d-galactopyranosides.
Catalog No. 50-290-1864 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10105031001
-Galactosidase or -D-Galactoside galactohydrolase is basically a tetramer made up of identical four polypeptide chains each constituting 1023 amino acids. It is very specific for D-galactose and requires K or Na and Mg2 to be fully active. -Galactosidase enzyme with an oxygen glycosidic bond catalyzes reactions with -d-galactopyranosides.