Learn More
Medchemexpress LLC Beta-galactosidase/GLB1 protein, human (HEK293, His tag) | 9031-11-2 | >95.0% | 100UG
SDP
Catalog No. 5000175820
Encompass_Preferred
Recombinant human beta-galactosidase (GLB1) is a purified, His-tagged enzyme produced in HEK293 cells for biochemical and cell biology research. It hydrolyzes beta-linked terminal galactose residues and is supplied as a filtered protein solution suitable for activity assays and antibody validation.
  • Human GLB1 expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • >95.0% purity by reducing SDS-PAGE.
  • Approximate molecular weight 90.0 kDa.
  • Supplied in 20 mM Tris-HCl, 150 mM NaCl, pH 8.0.
  • Shipped on dry ice and stored at -80°C for long-term stability.
  • Available in multiple sizes including 100 μg.
  • Suitable for enzyme activity assays, structural analysis, and antibody validation.
Catalog No. 50-001-75820 Supplier Medchemexpress LLC Supplier No. HYP70303100UG
Recombinant human beta-galactosidase (GLB1) is a purified, His-tagged enzyme produced in HEK293 cells for biochemical and cell biology research. It hydrolyzes beta-linked terminal galactose residues and is supplied as a filtered protein solution suitable for activity assays and antibody validation.
  • Human GLB1 expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • >95.0% purity by reducing SDS-PAGE.
  • Approximate molecular weight 90.0 kDa.
  • Supplied in 20 mM Tris-HCl, 150 mM NaCl, pH 8.0.
  • Shipped on dry ice and stored at -80°C for long-term stability.
  • Available in multiple sizes including 100 μg.
  • Suitable for enzyme activity assays, structural analysis, and antibody validation.