Learn More
Sigma Aldrich Fine Chemicals Biosciences beta-Glucanase from Tricho100G
Catalog No. 502900317
Encompass
-Glucanase represents a group of carbohydrate enzymes which break down glycosidic bonds within beta-glucan. It forms the main constituent of fungal cell walls and could be a potential structural and storage polysaccharide of marine macro-algae. It has the ability to degrade fungal cell walls and may be involved in defense mechanism of plants against pathogenic fungi.
Catalog No. 50-290-0317 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G4423100G
-Glucanase represents a group of carbohydrate enzymes which break down glycosidic bonds within beta-glucan. It forms the main constituent of fungal cell walls and could be a potential structural and storage polysaccharide of marine macro-algae. It has the ability to degrade fungal cell walls and may be involved in defense mechanism of plants against pathogenic fungi.