Learn More
Creative Biomart Beta-Glucocerebrosidase (GBA)
Supplier:  Creative Biomart THP0246
Encompass_Preferred
GBA produced in Sf9 Baculovirus cells is a single, glycosylated polypeptide chain containing 503 amino acids (40-536a.a.) and having a molecular mass of 56.4kDa. (Molecular size on SDS-PAGE will appear at approximately 50-70kDa). GBA is expressed with a 6 amino acid His tag at C-Terminus and purified by proprietary chromatographic techniques.
Catalog No. 50-208-4297