Learn More
TARGETMOL CHEMICALS INC beta-Glucogallin 5MG
SDP
Catalog No. 502879573
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. beta-Glucogallin (beta-Glucogallin) is a selective aldose reductase (AKR1B1) inhibitor with antioxidant, anti-glycation, and anti-inflammatory properties. beta-Glucogallin is a naturally occurring compound derived from Emblica officinalis and is used in diabetes research. Vitamin B3-d4 Purity 99.14%
Catalog No. 50-287-9573 Supplier TARGETMOL CHEMICALS INC Supplier No. T1260225MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. beta-Glucogallin (beta-Glucogallin) is a selective aldose reductase (AKR1B1) inhibitor with antioxidant, anti-glycation, and anti-inflammatory properties. beta-Glucogallin is a naturally occurring compound derived from Emblica officinalis and is used in diabetes research. Vitamin B3-d4 Purity 99.14%