Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Glucosidase from almo
Catalog No. 502907940
Encompass
-Glucosidase from almonds belongs to the family 1 of the glycoside hydrolases. Almonds contain prunasin hydrolases (PH) and amygdalin hydrolase. Nine types of PHs are associated with almonds. PHs have signal peptide sequence and correspond to molecular weight in the range 50 to 75 kDa. PHs also have ITENG NEP and INKKGIEYY motifs conserved and have N-glycosylation sites.
Catalog No. 50-290-7940 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G45111KU
-Glucosidase from almonds belongs to the family 1 of the glycoside hydrolases. Almonds contain prunasin hydrolases (PH) and amygdalin hydrolase. Nine types of PHs are associated with almonds. PHs have signal peptide sequence and correspond to molecular weight in the range 50 to 75 kDa. PHs also have ITENG NEP and INKKGIEYY motifs conserved and have N-glycosylation sites.