Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta Glucuronidase Antibody (YA2853) | 100 UL
SDP

Catalog No. 5000444420
Encompass_Preferred

Beta Glucuronidase Antibody (YA2853) from MedChemExpress is a rabbit-derived monoclonal antibody identified for research use. This antibody targets beta Glucuronidase and is suitable for various applications, including Western Blotting (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC).

  • Rabbit host
  • Human reactivity
  • Monoclonal antibody (YA2853)
  • Liquid appearance
  • Concentration of 1.5 mg/mL
  • Non-conjugated
  • Formulated in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles

Catalog No. 50-004-44420 Supplier Medchemexpress LLC Supplier No. HYP83108100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta Glucuronidase Antibody (YA2853) from MedChemExpress is a rabbit-derived monoclonal antibody identified for research use. This antibody targets beta Glucuronidase and is suitable for various applications, including Western Blotting (WB), Immunohistochemistry-Paraffin (IHC-P), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC).

  • Rabbit host
  • Human reactivity
  • Monoclonal antibody (YA2853)
  • Liquid appearance
  • Concentration of 1.5 mg/mL
  • Non-conjugated
  • Formulated in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.