Learn More
Medchemexpress LLC Beta-glucuronidase (bovine liver) | 9001-45-0 | 100 KU
SDP
Catalog No. 5000399531
Encompass_Preferred
Beta-glucuronidase (bovine liver) is a glycosyl hydrolase that facilitates the hydrolysis of β-glucuronic acid and sulfate esters found in various biological fluids, including urine. This enzymatic action results in the release of β-glucuronate.
  • Hydrolyzes β-glucuronic acid
  • Facilitates hydrolysis of sulfate esters
  • Derived from bovine liver
  • Functions within biological fluids
Catalog No. 50-003-99531 Supplier Medchemexpress LLC Supplier No. HYP2803A100KU
Beta-glucuronidase (bovine liver) is a glycosyl hydrolase that facilitates the hydrolysis of β-glucuronic acid and sulfate esters found in various biological fluids, including urine. This enzymatic action results in the release of β-glucuronate.
  • Hydrolyzes β-glucuronic acid
  • Facilitates hydrolysis of sulfate esters
  • Derived from bovine liver
  • Functions within biological fluids