Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-glucuronidase (Escherichia coli) | 9001-45-0 | ~290 kDa (tetramer), 68 kDa (monomer) | 25 KU
SDP

Catalog No. 5000397214
Encompass_Preferred

Beta-glucuronidase is a glycosyl hydrolase that hydrolyzes β-glucuronic acid and sulfate esters in urine and other biological fluids, releasing β-glucuronate. Its deficiency leads to mucopolysaccharidosis type VII (MPSVII), causing lysosomal storage in the brain. This product is for research use only.

  • Hydrolyzes β-glucuronic acid and sulfate esters
  • Optimum pH range of 6.0 to 7.0
  • Specific activity of 1,000,000-5,000,000 U/mg protein
  • Soluble in water at 10 mg/mL (needs ultrasonic and warming)
  • Unit definition: One unit liberates 1.0 microgram of phenolphthalein from phenolphthalein glucuronide per hour at pH 6.8 at 37°C (30 minute assay).

Catalog No. 50-003-97214 Supplier Medchemexpress LLC Supplier No. 25KUHYP2803
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Beta-glucuronidase is a glycosyl hydrolase that hydrolyzes β-glucuronic acid and sulfate esters in urine and other biological fluids, releasing β-glucuronate. Its deficiency leads to mucopolysaccharidosis type VII (MPSVII), causing lysosomal storage in the brain. This product is for research use only.

  • Hydrolyzes β-glucuronic acid and sulfate esters
  • Optimum pH range of 6.0 to 7.0
  • Specific activity of 1,000,000-5,000,000 U/mg protein
  • Soluble in water at 10 mg/mL (needs ultrasonic and warming)
  • Unit definition: One unit liberates 1.0 microgram of phenolphthalein from phenolphthalein glucuronide per hour at pH 6.8 at 37°C (30 minute assay).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.