Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Glucuronidase from Es
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G0799100KU
Encompass
-Glucuronidase is a tetramer belonging to the glycosidase family with a molecular weight of 29 kDa. It is composed of 74 kDa identical monomers. The pI and optimal pH of the enzyme is 4.8 and 6-7 respectively.
Catalog No. 50-290-2284