Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Glucuronidase from Es
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G8162100KU
Encompass
-Glucuronidase from Escherichia coli is similar to human glucuronidase enzyme and corresponds to molecular weight close to 69-71 kDa and has an pH optimum of 6.5-7.5. It belongs to family-2 glycosyl hydrolase and has active site residues glutamic acid 394 tyrosine 468 and glutamic acid 504.
Catalog No. 50-290-7341