Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Glucuronidase from He
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G07512MU
Encompass
beta-Glucuronidase Type H-2 from Helix pomatia is a crude SOLUTIONon of enzymes derived from the Roman snail. Many beta-glucuronidases derived from mollusks also contain sulfatase activity.
Catalog No. 50-290-2568