Learn More
Sigma Aldrich Fine Chemicals Biosciences beta Glucuronidase from He
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G701710ML
Encompass
beta-Glucuronidase Type H-2 from Helix pomatia is a crude SOLUTIONon of enzymes derived from the digestive juices of the Roman snail. beta-glucuronidases derived from mollusks often contain sulfatase activity. Both activities are useful in the hydrolysis of metabolites for toxicology and biomarker analysis. for a broader look at our beta-Glucuronidase product line and their uses see our page about Drug Metabolite Analysis
Catalog No. 50-290-2143