Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Beta-glucuronidase | 9001-45-0 | MFCD00199013 | >95.0% | 25UG
SDP

Catalog No. 5000176501
Encompass_Preferred

Recombinant beta-glucuronidase (GUSB) produced in Escherichia coli and supplied as an N-terminal 6xHis-tagged soluble enzyme. The preparation is provided as a filtered, stabilized solution suitable for biochemical assays and substrate hydrolysis studies using PNPG and 4-methylumbelliferyl-glucuronide.

  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight 69-78 kDa (full-length, M1-Q603).
  • Active with PNPG and 4-methylumbelliferyl-glucuronide substrates.
  • Expressed in Escherichia coli and supplied as a soluble protein.
  • N-terminal 6xHis tag for detection and affinity purification.
  • Formulated as a 0.2 μm filtered solution with buffers and stabilizers (trehalose, mannitol, Tween-80).
  • Available in small research quantities (for example, 25 μg).

Catalog No. 50-001-76501 Supplier Medchemexpress LLC Supplier No. HYP7025825UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant beta-glucuronidase (GUSB) produced in Escherichia coli and supplied as an N-terminal 6xHis-tagged soluble enzyme. The preparation is provided as a filtered, stabilized solution suitable for biochemical assays and substrate hydrolysis studies using PNPG and 4-methylumbelliferyl-glucuronide.

  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Molecular weight 69-78 kDa (full-length, M1-Q603).
  • Active with PNPG and 4-methylumbelliferyl-glucuronide substrates.
  • Expressed in Escherichia coli and supplied as a soluble protein.
  • N-terminal 6xHis tag for detection and affinity purification.
  • Formulated as a 0.2 μm filtered solution with buffers and stabilizers (trehalose, mannitol, Tween-80).
  • Available in small research quantities (for example, 25 μg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.