Learn More
ACROBiosystems beta-Glucuronidase/GUSB 100ug
Catalog No. 502535474
Encompass_Preferred
Human beta-Glucuronidase Protein His Tag (BEB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Thr 651 (Accession # P08236).
Catalog No. 50-253-5474 Supplier ACROBiosystems Supplier No. BEBH52H3100UG
Human beta-Glucuronidase Protein His Tag (BEB-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Thr 651 (Accession # P08236).