Learn More
Medchemexpress LLC Beta-glucuronidase (helix pomatia) | 9001-45-0 | 100 KU
SDP
Catalog No. 5000399532
Encompass_Preferred
Beta-glucuronidase (helix pomatia) is a glycosyl hydrolase that hydrolyzes β-glucuronic acid and sulfate esters in urine and other biological fluids, releasing β-glucuronate.
  • Functions as a glycosyl hydrolase
  • Hydrolyzes β-glucuronic acid
  • Hydrolyzes sulfate esters
  • Active in urine and other biological fluids
  • Releases β-glucuronate
Catalog No. 50-003-99532 Supplier Medchemexpress LLC Supplier No. HYP2803B100KU
Beta-glucuronidase (helix pomatia) is a glycosyl hydrolase that hydrolyzes β-glucuronic acid and sulfate esters in urine and other biological fluids, releasing β-glucuronate.
  • Functions as a glycosyl hydrolase
  • Hydrolyzes β-glucuronic acid
  • Hydrolyzes sulfate esters
  • Active in urine and other biological fluids
  • Releases β-glucuronate