Learn More
Medchemexpress LLC Beta-glucuronidase (helix pomatia) | 9001-45-0 | 50 KU
SDP
Catalog No. 5000396757
Encompass_Preferred
Beta-glucuronidase (helix pomatia) is a glycosyl hydrolase that hydrolyzes β-glucuronic acid and sulfate esters in urine and other biological fluids, and then releases β-glucuronate. It is for research use only.
  • Glycosyl hydrolase
  • Hydrolyzes β-glucuronic acid and sulfate esters
  • Releases β-glucuronate
  • Appearance: white to off-white solid
  • Store at -20°C for 2 years in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and sources of ignition
  • For research use only
  • Not fully validated for medical applications
Catalog No. 50-003-96757 Supplier Medchemexpress LLC Supplier No. HYP2803B50KU
Beta-glucuronidase (helix pomatia) is a glycosyl hydrolase that hydrolyzes β-glucuronic acid and sulfate esters in urine and other biological fluids, and then releases β-glucuronate. It is for research use only.
  • Glycosyl hydrolase
  • Hydrolyzes β-glucuronic acid and sulfate esters
  • Releases β-glucuronate
  • Appearance: white to off-white solid
  • Store at -20°C for 2 years in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and sources of ignition
  • For research use only
  • Not fully validated for medical applications